Learn More
Sino Biological Anti-KRT2/Cytokeratin2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105446T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human KRT2 / Cytokeratin2, and purified by antigen affinity chromatography.
Catalog No. 50-241-2306