Learn More
Sino Biological Human KRT2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 110167T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human KRT2/Cytokeratin2, and purified by antigen affinity chromatography.
Catalog No. 50-237-3594