Learn More
Sino Biological Anti-keratin 76/KRT76 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104712T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human KRT76, and purified by antigen affinity chromatography.
Catalog No. 50-240-8213