Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KRT77 (aa 24-60) Control Fragment Recombinant Protein

Catalog No. RP97883
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97883 Supplier Invitrogen™ Supplier No. RP97883
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (49%), Rat (49%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61130. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. KRT77 encodes an epithelial keratin that is expressed in the skin and eccrine sweat glands. The type II keratins are clustered in a region of chromosome 12q13.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7Z794
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 374454
Name Human KRT77 (aa 24-60) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 4732484G22Rik; CK-1B; CK-2P; cytokeratin 2; cytokeratin-1B; Cytokeratin-2P; embryonic type II keratin 1; embryonic type II keratin-1; HUMCYT2A; K1B; K1emb; K2P; K76; K77; Kb39; keratin 1b; keratin 2p; keratin 76; keratin 76, type II; keratin 77; keratin 77, type II; keratin complex 2, basic, gene 39; keratin, type II cytoskeletal 1b; Keratin, type II cytoskeletal 2 oral; keratin-76; Keratin-77; Krt1b; Krt239; KRT2B; KRT2P; KRT76; Krt77; mK1b; type II keratin Kb39; type-II intermediate filament protein; type-II keratin Kb39; Type-II keratin Kb9
Common Name KRT77
Gene Symbol KRT77
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SAGSGGGSPAVGSVCYARGRCGGGGYGIHGRGFGSRS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.