Learn More
Biomatik Corporation Human Kruppel Like Factor 5, Intestinal (KLF5) ELISA Kit
Supplier:  Biomatik Corporation EKU05520
Encompass_Preferred
Human Kruppel Like Factor 5, Intestinal (KLF5) ELISA Kit for detecting KLF5 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13887. Gene ID: 688. Target Synonyms: KLF5; BTEB2; CKLF; IKLF; Intestinal Enriched Krüppel-Like Factor; Colon krueppel-like factor; Basic Transcription Element Binding Protein 2; GC-box-binding protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5077