Learn More
Bioworld HUMAN KU P70/P80 (RECOMBINANT)
Supplier:  Bioworld 220603991
Encompass_Preferred
Recombinant Human Ku Protein(P70/P80) Source: Sf9 insect cells. Background: Ku antigen consists of two polypeptides (70 kDa and 80 kDa; the p80 subunit is occasionally called p86). The Ku heterodimer provides DNA-binding and regulato...
Catalog No. 50-253-496