Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KV4.3 (KCND3) (aa 527-653) Control Fragment Recombinant Protein

Catalog No. RP91276
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91276 Supplier Invitrogen™ Supplier No. RP91276
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56151. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shal-related subfamily, members of which form voltage-activated A-type potassium ion channels and are prominent in the repolarization phase of the action potential. This member includes two isoforms with different sizes, which are encoded by alternatively spliced transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UK17
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 3752
Name Human KV4.3 (KCND3) (aa 527-653) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AW045978; BRGDA9; KCND3; KCND3L; KCND3S; Kncd3; KSHIVB; KV4.3; potassium channel alpha subunit Kv4.3 short form; potassium channel Kv4.3; potassium channel Kv4.3L; potassium channel Kv4.3M; potassium channel, voltage gated Shal related subfamily D, member 3; potassium channel, voltage-gated Shal-related subfamily D, member 3; potassium ionic channel Kv4.3; potassium voltage gated channel, Shal-related family, member 3; potassium voltage-gated channel long; potassium voltage-gated channel subfamily D member 3; potassium voltage-gated channel, Shal-related family, member 3; potassium voltage-gated channel, Shal-related subfamily, member 3; SCA19; SCA22; sha1-related potassium channel Kv4.3; voltage-gated K+ channel; voltage-gated potassium channel subunit Kv4.3
Common Name KV4.3 (KCND3)
Gene Symbol KCND3
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NYPSTRSPSLSSHPGLTTTCCSRRSKKTTHLPNSNLPATRLRSMQELSTIHIQGSEQPSLTTSRSSLNLKADDGLRPNCKTSQITTAIISIPTPPALTPEGESRPPPASPGPNTNIPSIASNVVKVS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.