Learn More
Sino Biological Human L-selectin/CD62L Matched ELISA Antibody Pair Set 5 Plates
Supplier: Sino Biological SEK118385
The Human CD62L / L-Selectin / SELL ELISA Pair Set is for the quantitative determination of Human CD62L / L-Selectin / SELL.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CD62L / L-Selectin / SELL coated on a 96-well plate. Standards and samples are added to the wells, and any Human CD62L / L-Selectin / SELL present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human CD62L / L-Selectin / SELL monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CD62L / L-Selectin / SELL present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.