Learn More
Raybiotech Inc L-Series Human Obesity Antibody Array 182 Glass Slide Kit.
Catalog No. 501948498
Encompass_Preferred
Detects 182 Human Adipokines. Suitable for serum, plasma and cell culture supernatants. Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAH-BLG-ADI-1-4
Catalog No. 50-194-8498 Supplier Raybiotech Inc Supplier No. AAHBLGADI14
Detects 182 Human Adipokines. Suitable for serum, plasma and cell culture supernatants. Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAH-BLG-ADI-1-4