Learn More
Sino Biological Recombinant Human L1CAM Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10140H49HB100
Encompass_Preferred
A DNA sequence encoding the Human L1CAM (NP_000416.1) (Met1-Glu1120) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-0177