Learn More
Sino Biological Recombinant Human L1CAM Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10140H02H1
Encompass_Preferred
A DNA sequence encoding the Human L1CAM (NP_000416.1) (Met1-Glu1120) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-241-1619