Learn More
Raybiotech Inc L-Series Human Antibody Array 2000 Glass Slide Kit.
Supplier:  Raybiotech Inc AAHBLG20008
Encompass_Preferred
Detects 2000 Human Proteins. Suitable for serum, plasma and cell culture supernatants. (4 or 8 Sample Kit) Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAH-BLG-2000-8
Catalog No. 50-194-8494