Learn More
Biomatik Corporation Human Lactate Dehydrogenase D (LDHD) ELISA Kit
Supplier:  Biomatik Corporation EKU05536
Encompass_Preferred
Human Lactate Dehydrogenase D (LDHD) ELISA Kit for detecting LDHD in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86WU2. Gene ID: 197257. Target Synonyms: LDH-D; DLD; Probable D-lactate dehydrogenase, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6798