Learn More
Biomatik Corporation Human Lactoferrin (LTF) CLIA Kit
Supplier:  Biomatik Corporation EKU08915
Encompass_Preferred
Human Lactoferrin (LTF) CLIA Kit for detecting LTF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, breast milk, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 54.9-40,000pg/ml. Sensitivity: 23.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02788. Gene ID: 4057. Target Synonyms: HLF2; LF; GIG12; Lactotransferrin; Growth-inhibiting protein 12; Talalactoferrin; Lfcin-H; Kaliocin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7068