Learn More
ACROBiosystems Human LAG-3 25ug
Supplier:  ACROBiosystems LA3H82F325UG
Encompass_Preferred
Biotinylated Human LAG-3 Mouse IgG2a FcAvitag (LA3-H82F3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1).
Catalog No. 50-253-4526