Learn More
Bon Opus Biosciences Recombinant Human LAG3 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP101650UG
Encompass_Preferred
Recombinant Human LAG3 (C-mFc) is produced by Human Cells. The target gene encoding Leu23-Leu450 is expressed with a C-mFc tag. Accession# P18627. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1016
Catalog No. 50-217-2618