Learn More
Sino Biological Anti-LAG-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 16498T20100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LAG3. LAG3 specific IgG was purified by Human LAG3 affinity chromatography.
Catalog No. 50-237-1780