Learn More
ACROBiosystems ACROBiosystems Human LAIR-1 Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems LA1H5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human LAIR-1, Mouse IgG2a Fc Tag (LA1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GTX8-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 42.3 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Human LAIR-1, Mouse IgG2a Fc Tag (Cat. No. LA1-H5253) with a linear range of 10-78 ng/mL (QC tested).
Catalog No. 50-210-7103