Learn More
ACROBiosystems ACROBiosystems Human LAIR1 / CD305 Protein, His Tag
Catalog No. 502106562
Encompass_Preferred
Recombinant Protein;100UG;Human LAIR-1, His Tag (CD5-H52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GTX8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.4 kDa. The protein migrates as 26-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6562 Supplier ACROBiosystems Supplier No. CD5H52H1100UG
Recombinant Protein;100UG;Human LAIR-1, His Tag (CD5-H52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GTX8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.4 kDa. The protein migrates as 26-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.