Learn More
Sino Biological Recombinant Human LAIR1 Protein (ECD, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 13257H27HB100
Encompass_Preferred
A DNA sequence encoding the human LAIR1 (AAF71275.2) (Met1-Tyr165) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4262