Learn More
Abcam Human LAIR2 ELISA Kit, 96 Tests
Catalog No. 502712123
Encompass_Preferred
Human LAIR2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human LAIR2 with a sensitivity of 2.628 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-2123 Supplier Abcam Supplier No. AB31625196TESTS
Human LAIR2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human LAIR2 with a sensitivity of 2.628 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement