Learn More
Sino Biological Human LAIR2 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785617
Encompass_Preferred
The Human LAIR2 ELISA Pair Set is for the quantitative determination of Human LAIR2.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human LAIR2 coated on a 96-well plate. Standards and samples are added to the wells, and any Human LAIR2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human LAIR2 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human LAIR2 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85617 Supplier Sino Biological Supplier No. SEK108685
The Human LAIR2 ELISA Pair Set is for the quantitative determination of Human LAIR2.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human LAIR2 coated on a 96-well plate. Standards and samples are added to the wells, and any Human LAIR2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human LAIR2 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human LAIR2 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.