Learn More
Genscript Corporation Human Lambda Light Chain 2D54
Supplier: Genscript Corporation V069010.1
Immunoglobulins (Ig) and free light chains (FLC) is involved in humoral immune response Ig is composed of two identical light chains and two identical heavy chains FLC contains two types kappa free light chain (KFLC) and lambda free light chain (LFLC) The expression of Kappa or Lambda is greatly increased in multiple myeloma or other B cell malignancies It is useful for the identification of certain non-Hodgkin s lymphomas leukemias and plasmacytomas Immunohistochemistry is commonly applied for the detection of free light chains