Learn More
Abcam Human Lambda Light chain Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502689367
Encompass_Preferred
Human Lambda Light chain Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Lambda Light chain.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-9367 Supplier Abcam Supplier No. AB30854710X96TESTS
Human Lambda Light chain Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Lambda Light chain.
The product is subject to the following: Abcam Restricted Use Statement