Learn More
ACROBiosystems Human Laminin 521 Protein, premium grade, 100ug
Supplier:  ACROBiosystems LA5H5261100UG
Encompass_Preferred
Laminin 521 (LN521) is a recombinant human protein that provides a defined surface for in vitro feeder-free culture of multiple human pluripotent stem cells (PSCs). LN521 has been proven to maintain normal growth characteristics and stemness in multiple PSC lines without simultaneous differentiation, which includes ESC, iPSC, MSC etc. In addition, LN521 has been demonstrated to support PSC growth for >10 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages.
Catalog No. 50-233-0916