Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Laminin alpha 4 ELISA Kit

Catalog No. EH293RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH293RB 96 Tests
EH293RBX5 5 x 96 Tests
EH293RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH293RB Supplier Invitrogen™ Supplier No. EH293RB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human Laminin alpha 4 quantitates human Laminin alpha 4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Laminin alpha 4.

Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16363
Assay Range 1.63-400ng/mL
Assay Sensitivity 1.63 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CMD1JJ; LAMA3; LAMA4; LAMA4*-1; laminin [a]4; laminin alpha 4 chain; laminin subunit alpha 4; laminin subunit alpha-4; laminin, alpha 4; laminin-14 subunit alpha; Laminin-8 subunit alpha; laminin-9 subunit alpha; RGD1560062CMD1JJ; LAMA3; LAMA4; LAMA4*-1; laminin [a]4; laminin alpha 4 chain; laminin subunit alpha 4; laminin subunit alpha-4; laminin, alpha 4; laminin-14 subunit alpha; Laminin-8 subunit alpha; laminin-9 subunit alpha; RGD1560062;
Gene ID (Entrez) 3910
Gene Symbol LAMA4
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Laminin alpha-4
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.