Learn More
Biomatik Corporation Human Laminin Beta 2 (LAMb2) ELISA Kit
Supplier:  Biomatik Corporation EKU05559
Encompass_Preferred
Human Laminin Beta 2 (LAMb2) ELISA Kit for detecting LAMb2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55268. Gene ID: 3913. Target Synonyms: LAM-B2; LAMS; Laminin S; Laminin B1s chain; S-laminin subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7108