Learn More
Biomatik Corporation Human Laminin Beta 3 (LAMb3) ELISA Kit
Supplier:  Biomatik Corporation EKU05560
Encompass_Preferred
Human Laminin Beta 3 (LAMb3) ELISA Kit for detecting LAMb3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13751. Gene ID: 3914. Target Synonyms: LAM-B3; LAMNB1; Nicein subunit beta; Kalinin subunit beta; Epiligrin subunit bata; Laminin B1k chain; Laminin-5 subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8897