Learn More
Biomatik Corporation Human Laminin (LN) CLIA Kit
Supplier:  Biomatik Corporation EKU09425
Encompass_Preferred
Human Laminin (LN) CLIA Kit for detecting LN in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.9-5,000pg/ml. Sensitivity: 2.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P25391; P07942; P11047. Gene ID: 284217; 3912; 3915. Target Synonyms: LAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8522