Learn More
Biomatik Corporation Human Laminin (LN) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10293
Encompass_Preferred
Human Laminin (LN) High-sensitive ELISA Kit for detecting LN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25391; P07942; P11047. Gene ID: 3912; 3915; 284217. Target Synonyms: LAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7287