Learn More
R&D Systems™ Human LAMP-2B/CD107b (NP_054701) VersaClone cDNA
Catalog No. RDC1003
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC1003 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC1003 Supplier R&D Systems™ Supplier No. RDC1003
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lysosomal-associated membrane protein 2
The Human LAMP-2B/CD107b (NP_054701) VersaClone cDNA clone contains the complete ORF for LAMP-2/CD107b, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LAMP-2/CD107b insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Gene Alias CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP2, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein 2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans