Learn More
Sino Biological Recombinant Anti-LAMP1 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 11215R107A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human LAMP-1 / CD107a (rh LAMP-1 / CD107a; Catalog#11215-H08H; NP_005552.3; Met1-Met382) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-0657