Learn More
Sino Biological Anti-LAMP1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11215MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LAMP-1 / CD107a / LAMP1 (rh LAMP-1 / CD107a / LAMP1; Catalog#11215-H08H; NP_005552.3; Met1-Met382). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1284