Learn More
Sino Biological Anti-LAMP2 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 13555MM05F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LAMP2/CD107b (rh LAMP2/CD107b; Catalog#13555-H08H; NP_054701.1; Met1-Ile375) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-255-4216