Learn More
Sino Biological Anti-LAMP2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13555MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LAMP2/CD107b (rh LAMP2/CD107b; Catalog#13555-H08H; NP_054701.1; Met1-Ile375). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4214