Learn More
Sino Biological Anti-LAMP3/CD208 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10527MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD208 / LAMP3 (rh LAMP3; Catalog#10527-H08H; NP_055213.2; Lys 28-Thr 381). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1838