Learn More
Sino Biological Recombinant Anti-LAMP3/CD208 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10527R00220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD208 / LAMP3 (rh LAMP3; Catalog#10527-H08H; NP_055213.2; Lys 28-Thr 381).
Catalog No. 50-241-1874