Learn More
Sino Biological Recombinant Anti-LAMP3/CD208 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10527R02020
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD208 / DC-LAMP (rh CD208 / DC-LAMP; Catalog#10527-H08H; NP_055213.2; Lys28-Thr381).
Catalog No. 50-240-8249