Learn More
Sino Biological Anti-LAMP3/CD208 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10527T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LAMP3 (rh LAMP3; Catalog#10527-H08H; NP_055213.2; Lys28-Thr381). LAMP3 specific IgG was purified by Human LAMP3 affinity chromatography.
Catalog No. 50-241-0856