Learn More
R&D Systems™ Human LAP (TGF-beta 1) Quantikine ELISA Kit

Description
Specifications
Specifications
| Assay Range | 23.40 - 1500pg/mL |
| Assay Sensitivity | 3.4pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Platelet-poor Plasma (EDTA, Heparin), Serum, Urine |
| Specificity | Natural and Recombinant Human LAP (TGF-beta 1) and Latent TGF-beta 1). |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7040 |
| Sample Volume | Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (10uL),Platelet-poor EDTA Plasma (10uL),Platelet-poor Heparin Plasma (10uL),Urine (50uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.