Learn More
Sino Biological Anti-LAP3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 15480RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LAP3 (rh LAP3; Catalog#15480-H08H; NP_056991.2; Met1-Ala519). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4099