Learn More
BIOHIPPO Human LAT Microsample
SDP
Catalog No. 5000314220
Encompass
Human LAT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LAT in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 109 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LAT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LAT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LAT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14220 Supplier BIOHIPPO Supplier No. BHE15212125
Human LAT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LAT in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 109 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LAT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LAT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LAT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color