Learn More
ACROBiosystems ACROBiosystems Human Latent GDF-2 / BMP-9 Protein, His Tag (MALS verified)
Catalog No. 502106334
Encompass_Preferred
Recombinant Protein;50UG;Human Latent GDF-2, His Tag (GD2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 23 - Arg 429 (Accession # Q9UK05-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 47.0 kDa. The protein migrates as 13 kDa and 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6334 Supplier ACROBiosystems Supplier No. GD2H52H350UG
Recombinant Protein;50UG;Human Latent GDF-2, His Tag (GD2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 23 - Arg 429 (Accession # Q9UK05-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 47.0 kDa. The protein migrates as 13 kDa and 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.