Learn More
ACROBiosystems ACROBiosystems Human latent GDF-8 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems GD8H5243500UG
Encompass_Preferred
Recombinant Protein;500UG;Human latent GDF-8, His Tag (GD8-H5243) is expressed from human 293 cells (HEK293). It contains AA Asn 24 - Ser 375 (Accession # O14793-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 42 kDa.
Catalog No. 50-210-6185