Learn More
ACROBiosystems ACROBiosystems Human Latent TGF-beta 3 / Latent TGFB3 Protein, Tag Free
Supplier:  ACROBiosystems TG3H521310UG
Encompass_Preferred
Recombinant Protein;10UG;Human Latent TGF-beta 3 / Latent TGFB3, Tag Free (TG3-H5213) is expressed from human 293 cells (HEK293). It contains AA Ala 301 - Ser 412 (Accession # P10600-1).;This protein carries no "tag". The protein has a calculated MW of 12.7 kDa. The protein migrates as 12 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human Latent TGF-beta 3 / Latent TGFB3, Tag Free (Cat. No. TG3-H5213) at 2 μg/mL (100 μL/well) can bind Biotinylated Human TGF-beta RII, His,Avitag (Cat. No. TG2-H82E4) with a linear range of 1-10 ng/mL (QC tested).
Catalog No. 50-210-7077