Learn More
American Research Products Inc Human latent transforming grow
Catalog No. 5000326054
Encompass
Human latent transforming growth factor beta binding protein 2 LTBP2 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26054 Supplier American Research Products Inc Supplier No. CSBE11268H
Human latent transforming growth factor beta binding protein 2 LTBP2 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich