Learn More
Sino Biological Human LAYN MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10208R10720
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Layilin (rh Layilin; Catalog#10208-H08H; NP_849156.1; Met 1-Glu 220).
Catalog No. 50-237-1523