Learn More
Proteintech Group Inc Human LBP ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0013496T
Encompass_Preferred
LBP (lipopolysaccharide-binding protein) is a serum glycoprotein belonging to the BPI/LBP/Plunc superfamily and BPI/LBP family. It is an acute-phase protein, predominantly synthesized by hepatocytes. LBP is present in normal serum at concentrations of 5 to 10 μg/ml, rising up to 200 μg/ml 24 h after induction of an acute-phase response. LBP binds to the lipid A moiety of bacterial lipopolysaccharides (LPS) which is a glycolipid present in the outer membrane of all Gram-negative bacteria, and acts as an affinity enhancer for CD14, facilitating its association with LPS.
Catalog No. 50-197-4093