Learn More
Sino Biological Human LCN1 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786359
Encompass_Preferred
The human LCN1 / Lipocalin 1 ELISA Pair Set is for the quantitative determinationof human LCN1 / Lipocalin 1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for LCN1 / Lipocalin 1 coated on a 96-well plate. Standards and samples are added to the wells, and any LCN1 / Lipocalin 1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-LCN1 / Lipocalin 1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of LCN1 / Lipocalin 1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86359 Supplier Sino Biological Supplier No. SEK1158315
The human LCN1 / Lipocalin 1 ELISA Pair Set is for the quantitative determinationof human LCN1 / Lipocalin 1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for LCN1 / Lipocalin 1 coated on a 96-well plate. Standards and samples are added to the wells, and any LCN1 / Lipocalin 1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-LCN1 / Lipocalin 1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of LCN1 / Lipocalin 1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.