Learn More
Sino Biological Recombinant Human LDLR/LDL Receptor Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10231H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human LDLR (NP_000518.1) precursor (Met 1-Arg 788) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6771